Skip to Content
Merck
All Photos(1)

Documents

96924

Sigma-Aldrich

2,2,2-Trifluoroethanol

BioUltra, for molecular biology, ≥99.0% (GC)

Synonym(s):

Trifluoroethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CH2OH
CAS Number:
Molecular Weight:
100.04
Beilstein:
1733203
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

vapor density

3.5 (vs air)

vapor pressure

70 mmHg ( 25 °C)

product line

BioUltra

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

~896 °F

expl. lim.

42 %

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

refractive index

n20/D 1.3 (lit.)

bp

77-80 °C (lit.)

mp

−44 °C (lit.)

density

1.373 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤1 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: ≤0.03
λ: 280 nm Amax: ≤0.02

SMILES string

OCC(F)(F)F

application(s)

sample preparation

InChI

1S/C2H3F3O/c3-2(4,5)1-6/h6H,1H2

InChI key

RHQDFWAXVIIEBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2,2-Trifluoroethanol (Trifluoroethyl alcohol, TFE) is used to study the conformational states of proteins and the folding refolding processes of proteins.

Other Notes

Solvent used for sample preparation in electrophoresis

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Shiraki et al.
Journal of molecular biology, 245(2), 180-194 (1995-01-13)
Trifluoroethanol (TFE) is known to stabilize the alpha-helical structure in proteins and their fragments. However, the relationship between the TFE-induced structures and the native structure is not clear. Here we show that beta-lactoglobulin, which consists predominantly of beta-sheets, exhibited a
T Rabilloud et al.
Electrophoresis, 19(6), 1006-1014 (1998-06-25)
Owing to the complexity of higher eukaryotic cells, characterization of a complete proteome is likely to be difficult to achieve. However, advantage can be taken of the cell compartmentalization to build organelle proteomes, which can moreover be viewed as specialized
Na Zhang et al.
Biomaterials science, 8(22), 6286-6300 (2020-10-07)
The regeneration of injured neurons over long injury distances remains suboptimal. In order to successfully stimulate nerve regrowth, potent biomolecules are necessary. Furthermore, reproducible and translatable methods to test the potency of candidate drugs for enhancing nerve regeneration over long
Gregory Cole et al.
Biochemistry, 51(25), 5033-5040 (2012-06-07)
Tetherin/Bst-2 is a cell surface protein that can act as a restriction factor against a number of enveloped viruses, including HIV-1. It acts by tethering new virus particles to the host cell membrane, promoting their internalization and degradation. Tetherin is
Bei Feng et al.
Biomacromolecules, 13(12), 3917-3925 (2012-11-08)
In tissue engineering research, there has recently been considerable interest in using electrospun biomimetic nanofibers of hybrids, in particular, from natural and synthetic polymers for engineering different tissues. However, phase separation between a pair of much dissimilar polymers might give

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service