Skip to Content
Merck
All Photos(1)

Documents

88821

Sigma-Aldrich

Atto Rho6G

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

product line

BioReagent

Assay

90.0% (HPLC)

form

solid

manufacturer/tradename

ATTO-TEC GmbH

transmittance

254 nm
535 nm

fluorescence

λex 535 nm; λem 560 nm

λ

in ethanol (with 0.1% trifluoroacetic acid)

UV absorption

λ: 532-538 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto Rho6G is a new rhodamine dye, based on the well-known laser dye Rhodamine 6G. The new label is functionalized for coupling to biomolecules such as DNA, RNA or proteins. Atto Rho6G shows strong absorption, extraordinary high fluorescence quantum yield, high thermal and photo-stability, and very little triplet formation.
After coupling to a substrate Atto Rho6G carries a net electric charge of +1.
The label is moderately hydrophilic

find more information here

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service