Skip to Content
Merck
All Photos(1)

Documents

86090201

HeLa229 cell line

from human cervix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human cervix

form

liquid

growth mode

Adherent

karyotype

2n = 46

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human cervix carcinoma

Cell Line Description

Derived from the parent HeLa line but differs chiefly in its relative insusceptibility to polioviruses. The presence of human papilloma virus 18 (HPV-18) sequences in HeLa cells has been reported.

Application

Tumourigenicity and virus studies, poliovirus 1 and AD3

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 9,10
D13S317: 14
D16S539: 9,10
D5S818: 11,12
D7S820: 8,12
THO1: 7
TPOX: 8,12
vWA: 16,18

Culture Medium

EMEM (HBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luis Crovetto et al.
The journal of physical chemistry. A, 119(44), 10854-10862 (2015-10-16)
Dyes with near-red emission are of great interest because of their undoubted advantages for use as probes in living cells. In-depth knowledge of their photophysics is essential for employment of such dyes. In this article, the photophysical behavior of a
C Yee et al.
The American journal of pathology, 119(3), 361-366 (1985-06-01)
A series of human carcinoma cell lines was examined for human papillomavirus (HPV) DNA sequences with the use of HPV-6, HPV-11, HPV-16, and HPV-18 DNA probes. Six of eight cell lines derived from human cervical carcinomas were shown to contain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service