Skip to Content
Merck
All Photos(1)

Key Documents

73021

Sigma-Aldrich

MES pH 6.5; 0.05M – DTT 0.001M Solution solution

Synonym(s):

Crystallization solution 05/Kit-No 96827

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

solution

reaction suitability

reagent type: reductant

pH

6.5±0.2 ( in neat)

density

1.00 g/mL at 20 °C

storage temp.

2-8°C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service