Skip to Content
Merck
All Photos(2)

Documents

62501-026

Sigma-Aldrich

DNAgard® Blood

10mL bottle, for room temp preservation of DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

for molecular biology

form

liquid

storage temp.

room temp

Packaging

1 bottle

Physical form

DNAgard Blood reagent 10 mL (stabilizes up to 40 mL of blood)

Legal Information

DNAgard is a registered trademark of Biomatrica

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malini Udtha et al.
Biopreservation and biobanking, 12(5), 332-336 (2014-10-24)
Preservation of biospecimens for biobanking applications traditionally involves freezing while maintaining the integrity of the product throughout multiple freeze-thaw cycles. The protection and stabilization of DNA at room temperature (RT) may eliminate the costs associated with freezer storage and reduce

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service