Skip to Content
Merck
All Photos(1)

Key Documents

42355

Sigma-Aldrich

DGEA

≥97% (TLC)

Synonym(s):

α2β1 Integrin Recognition Sequence, L-α-Aspartylglycyl-L-α-glutamyl-L-alanine, H-Asp-Gly-Glu-Ala-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N4O9
CAS Number:
Molecular Weight:
390.35
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

Assay

≥97% (TLC)

storage temp.

−20°C

SMILES string

C[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)CNC(=O)[C@@H](N)CC(O)=O)C(O)=O

InChI

1S/C14H22N4O9/c1-6(14(26)27)17-13(25)8(2-3-10(20)21)18-9(19)5-16-12(24)7(15)4-11(22)23/h6-8H,2-5,15H2,1H3,(H,16,24)(H,17,25)(H,18,19)(H,20,21)(H,22,23)(H,26,27)/t6-,7-,8-/m0/s1

InChI key

HZHXMUPSBUKRBW-FXQIFTODSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W D Staatz et al.
The Journal of biological chemistry, 266(12), 7363-7367 (1991-04-25)
The alpha 2 beta 1 integrin serves as either a specific cell surface receptor for collagen or as both a collagen and laminin receptor depending upon the cell type. Recently we established that the alpha 2 beta 1 integrin binds
N Z Gebrehiwot et al.
BMC genomics, 21(1), 869-869 (2020-12-09)
Humpless Bos taurus cattle are one of the earliest domestic cattle in Africa, followed by the arrival of humped Bos indicus cattle. The diverse indigenous cattle breeds of Africa are derived from these migrations, with most appearing to be hybrids
Jennifer L Bain et al.
Tissue engineering. Part A, 21(17-18), 2426-2436 (2015-07-16)
Autogenous bone is the gold standard material for bone grafting in craniofacial and orthopedic regenerative medicine. However, due to complications associated with harvesting donor bone, clinicians often use commercial graft materials that may lose their osteoinductivity due to processing. This
Cathy Nangini et al.
Scientific data, 6, 180280-180280 (2019-01-16)
We present a global dataset of anthropogenic carbon dioxide (CO2) emissions for 343 cities. The dataset builds upon data from CDP (187 cities, few in developing countries), the Bonn Center for Local Climate Action and Reporting (73 cities, mainly in
Akiko Niibori-Nambu et al.
PloS one, 8(5), e59558-e59558 (2013-05-25)
Glioma initiating cells (GICs) are considered responsible for the therapeutic resistance and recurrence of malignant glioma. To clarify the molecular mechanism of GIC maintenance/differentiation, we established GIC clones having the potential to differentiate into malignant gliomas, and subjected to DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service