Skip to Content
Merck
All Photos(1)

Key Documents

158240

Sigma-Aldrich

1-Nitrosopyrrolidine

99%

Synonym(s):

N-Nitrosopyrrolidine, NPYR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N2O
CAS Number:
Molecular Weight:
100.12
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

99%

refractive index

n20/D 1.489 (lit.)

bp

214 °C (lit.)

density

1.085 g/mL at 25 °C (lit.)

SMILES string

O=NN1CCCC1

InChI

1S/C4H8N2O/c7-5-6-3-1-2-4-6/h1-4H2

InChI key

WNYADZVDBIBLJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Nitrosopyrrolidine can be used to study carcinogens and cancer. Metabolism of nitrosamines in vivo has been tested in rats.1-Nitrosopyrrolidine has been used in a study to assess nitric oxide and spontaneous motility in chick embryos.

Biochem/physiol Actions

Induces hepatocellular carcinomas and lung adenomas in mice. Forms DNA adducts that primarily result in A:T to G:C mutations.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nuria Arranz et al.
Journal of applied toxicology : JAT, 26(5), 466-473 (2006-07-28)
The aim of this study was to investigate the protective effect of isothiocyanates towards N-nitrosamine-induced DNA damage in the single-cell gel electrophoresis (SCGE)/HepG2 assay. None of the isothiocyanates (ITCs) concentrations tested in the presence or absence of formamidopyrimidine-DNA glycosylase (Fpg)
Mingyao Wang et al.
Chemical research in toxicology, 20(4), 625-633 (2007-03-31)
N-Nitrosopyrrolidine (NPYR) is a well-established hepatocarcinogen in the rat. NPYR requires metabolic activation by cytochrome P450-catalyzed alpha-hydroxylation to express its carcinogenic activity. This produces alpha-hydroxyNPYR (2), which spontaneously ring opens to 4-oxobutanediazohydroxide (4), a highly reactive intermediate, which may itself
Nuria Arranz et al.
Journal of applied toxicology : JAT, 28(6), 788-796 (2008-03-18)
The aim of this study was to evaluate the effect of vitamin C towards N-nitrosopyrrolidine (NPYR)- and N-nitrosodimethylamine (NDMA)-induced apoptosis in human hepatoma (HepG2) and leukemia (HL-60) cell lines using flow cytometry analysis and the terminal deoxynucleotidyl transferase-mediated dUTP nick
F W Krüger et al.
Zeitschrift fur Krebsforschung und klinische Onkologie. Cancer research and clinical oncology, 83(3), 255-260 (1975-01-01)
After i. p. application of 6 mg/30 muCi/kg 2,5- and 3,4--14C-nitrosopyrrolidine about 20% of the radioactivity is exhaled as 14-CO2 and about 7% is found in the urine. One of the urinary metabolites was identified by thin layer chromatography, combined
Sadagopan Krishnan et al.
Chemical communications (Cambridge, England), (17)(17), 1713-1715 (2007-04-26)
We report for the first time voltammetric/electrochemiluminescent sensors applied to predict genotoxicity of N-nitroso compounds bioactivated by human cytochrome P450 enzymes.

Articles

DNA damage and repair mechanism is vital for maintaining DNA integrity. Damage to cellular DNA is involved in mutagenesis, the development of cancer among others.

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service