Skip to Content
Merck
All Photos(1)

Key Documents

13110

Sigma-Aldrich

N-Benzoyl-L-tyrosine ethyl ester

≥98.0% (NT)

Synonym(s):

BTEE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NHCOC6H5)CO2C2H5
CAS Number:
Molecular Weight:
313.35
Beilstein:
2223819
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:

Product Name

N-Benzoyl-L-tyrosine ethyl ester, ≥98.0% (NT)

Assay

≥98.0% (NT)

form

powder or crystals

optical activity

[α]20/D −26±3°, c = 1% in ethanol

color

white

mp

118-121 °C (lit.)
118-122 °C

solubility

methanol: 1 g/10 mL, clear, colorless to almost colorless (















)

SMILES string

CCOC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)c2ccccc2

InChI

1S/C18H19NO4/c1-2-23-18(22)16(12-13-8-10-15(20)11-9-13)19-17(21)14-6-4-3-5-7-14/h3-11,16,20H,2,12H2,1H3,(H,19,21)/t16-/m0/s1

InChI key

SRLROPAFMUDDRC-INIZCTEOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Benzoyl-L-tyrosine ethyl ester (BTEE) is a substrate which is completely resistant to trypsin and is required for the assay of chymotrypsin.[1]

Application

N-Benzoyl-L-tyrosine ethyl ester has been used for measuring chymotrypsin activity.[2][3]

Other Notes

Substrate, completely resistant to trypsin, for the assay of chymotrypsin[4][5][6]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Substrate solubilization for the Hummel alpha-chymotrypsin assay.
K N Rao et al.
Analytical biochemistry, 65(1-2), 548-551 (1975-05-12)
Hydrolysis of polyesters by serine proteases.
Lim HA
Biotechnology Letters, 27, 459-464 (2005)
J Kantorski et al.
Journal of bioluminescence and chemiluminescence, 7(1), 37-45 (1992-01-01)
We have studied an indirect role of serine and thiol proteases in the activation of human neutrophils in vitro. Stimulation was evaluated using a chemiluminescence (CL) generation system. Receptor-dependent and receptor-independent stimuli were studied, e.g. opsonized zymosan, formyl-methionyl-leucyl-phenylalanine, platelet activating
R Chatterjee et al.
Photochemistry and photobiology, 51(1), 91-97 (1990-01-01)
Skh:HR-1 hairless mice were irradiated chronically with sub-erythemal doses of UVB radiation, and a number of biochemical parameters in the skin were determined after 6, 12, 18, and 24 wk of exposure. The parameters measured were water, collagen, elastin, and
Effects of dietary essential oil components on growth performance, digestive enzymes and lipid metabolism in female broiler chickens.
Lee KW
British Poultry Science, 44, 450-450 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service