Skip to Content
Merck
All Photos(1)

Documents

11184

Sigma-Aldrich

N-Biotinyl-3,6-dioxaoctane-1,8-diamine trifluoroacetate salt solution

≥95% (TLC), 25 mg/mL in DMSO

Synonym(s):

N-{2-[2-(2-Aminoethoxy)ethoxy]ethyl}biotinamide trifluoroacetate salt, Biotin-DADOO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H30N4O4S · C2HF3O2
CAS Number:
Molecular Weight:
488.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Assay

≥95% (TLC)

concentration

25 mg/mL in DMSO

solubility

DMF: soluble

storage temp.

2-8°C

SMILES string

OC(=O)C(F)(F)F.NCCOCCOCCNC(=O)CCCC[C@@H]1SCC2NC(=O)NC12

InChI

1S/C16H30N4O4S.C2HF3O2/c17-5-7-23-9-10-24-8-6-18-14(21)4-2-1-3-13-15-12(11-25-13)19-16(22)20-15;3-2(4,5)1(6)7/h12-13,15H,1-11,17H2,(H,18,21)(H2,19,20,22);(H,6,7)/t12-,13-,15-;/m0./s1

InChI key

SJXUUAFYSXSFNA-HZPCBCDKSA-N

Application

Utilized to biotinylate thiomethyl and thiosulfate derivatives of carboxymethylcellulose through a DCC coupling reaction Labeling targets include glycoproteins, glycopeptides, and any aldehyde- or keto group-containing component (e.g., haptens) Utilized to synthesize a biotin-estradiol conjugate (i.e., biotinoyl-diaminodioxaoctane-estradiol derivative) to develop a direct, broad range enzyme immunoassay to measure plasma estradiol concentrations.

Packaging

Sold on the basis of grams of solution.
Product 11184-1G contains 1 gram of solution (approximately 910 microliters), at a concentration of 25 mg/mL (~23 mg N-Biotinyl-3,6-dioxaoctane-1,8-diamine trifluoroacetate salt).

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Miyatake et al.
Molecular and cellular biology, 11(12), 5894-5901 (1991-12-01)
The region extending from -40 to -54 of the 5'-flanking region of the mouse granulocyte-macrophage colony-stimulating factor (GM-CSF) gene shows homology to sequences found in the 5'-flanking regions of other cytokine genes, those encoding interleukin-4 (IL-4), IL-5, and granulocyte colony-stimulating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service