05801
D-Galactose 6-sulfate sodium salt
≥98.0% (TLC)
Synonym(s):
Gal-6S
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H11O9SNa
CAS Number:
Molecular Weight:
282.20
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
Recommended Products
Assay
≥98.0% (TLC)
solubility
water: soluble
storage temp.
−20°C
SMILES string
[Na+].OC1O[C@H](COS([O-])(=O)=O)[C@H](O)[C@H](O)[C@H]1O
InChI
1S/C6H12O9S.Na/c7-3-2(1-14-16(11,12)13)15-6(10)5(9)4(3)8;/h2-10H,1H2,(H,11,12,13);/q;+1/p-1/t2-,3+,4+,5-,6?;/m1./s1
InChI key
MKFZWUBBFWXXKZ-HLTNBIPTSA-M
Looking for similar products? Visit Product Comparison Guide
Application
D-Galactose 6-sulfate (Gal-6S) may be used to help differentiate glycan sulfatide recognition/binding sites and to study the structure and biochemisty of carrageenans and galactans.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Rosiane G M Zibetti et al.
Carbohydrate research, 344(17), 2364-2374 (2009-10-06)
Cryptonemia seminervis biosynthesizes a family of D,L-hybrid galactans based on the classical 3-linked beta-D-galactopyranosyl-->4-linked alpha-D- and alpha-L-galactopyranosyl alternating sequence (A-units-->B-units) with major amounts of alpha-D- and alpha-L-galactose and 3,6-anhydro-D- and L-galactose and lesser percentages of 3,6-anhydro-2-O-methyl-L-galactose, 2-O-methyl-, 4-O-methyl- and 6-O-methylgalactoses.
Carina Gonçalves et al.
FEMS microbiology letters, 281(1), 30-35 (2008-02-19)
Escherichia coli heat-STb is an important cause of diarrhea in piglets. STb was shown to interact specifically with sulfatide (3'-sulfogalactosyl-ceramide) present on the surface of epithelial cells of piglet jejunum. Basic data are lacking on STb binding to sulfatide in
Diego A Navarro et al.
Carbohydrate research, 338(20), 2111-2118 (2003-09-25)
A combination of two reported procedures was used in order to determine the configuration of the 3,6-anhydrogalactose present in red seaweed polysaccharides. A mild hydrolysis (to cleave only 3,6-anhydrogalactosyl linkages) was followed by a reductive amination with a chiral amine.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service