Skip to Content
Merck
All Photos(1)

Key Documents

379964

Supelco

Calcium acetate hydrate

99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Ca · xH2O
CAS Number:
Molecular Weight:
158.17 (anhydrous basis)
Beilstein:
3692527
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

99.99% trace metals basis

form

powder

impurities

<1% acetic acid

SMILES string

O.CC(=O)O[Ca]OC(C)=O

InChI

1S/2C2H4O2.Ca.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+2;/p-2

InChI key

XQKKWWCELHKGKB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium acetate hydrate is an organic calcium salt. It is the principal component of PA (pyroligneous acid)-modified limestone.[1]

Application

Calcium acetate hydrate may be used as the following:
  • Precursor to produce calcium ions in the development of CaO-based mesoporous CO2 sorbent.[2][3]
  • Precursor in the synthesis of Fe3O4 (magnetite )–Ca3(PO4)2 (calcium phosphate) core–shell nanoparticles.[4]
  • Raw material in the synthesis hydroxyapatite–wollastonite composites.[5]
  • Component of the reservoir solution used to induce crystallization of proteins.[6]
  • Crosslinking agent in the microfluidic production of alginate hydrogel microbeads.[7]
  • To rinse the glass surfaces before bonding during the formation of nano channels.[8]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nano-channel formation using a low-temperature glass-to-glass bonding process.
Schultz A, et al.
Proceedings of the Institution of Mechanical Engineers, Part N: Journal of Nanoengineering and Nanosystems, 224(4), 183-187 (2010)
Cyclic CO2 capture behavior of limestone modified with pyroligneous acid (PA) during calcium looping cycles.
Li Y, et al.
Industrial & Engineering Chemistry Research, 50(17), 10222-10228 (2011)
Investigating the optimum conditions for the formation of calcium oxide, used for CO2 sequestration, by thermal decomposition of calcium acetate.
Bilton M, et al.
Journal of Physics. Conference Series, 371(1) (2012)
Integrated parallel microfluidic device for simultaneous preparation of multiplex optical-encoded microbeads with distinct quantum dot barcodes.
Ji XH, et al.
Journal of Materials Chemistry, 21(35), 13380-13387 (2011)
Mechanical and bioactive behavior of hydroxyapatite-wollastonite sintered composites.
Encinas-Romero MA, et al.
International Journal of Applied Ceramic Technology, 7(2), 164-177 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service