Skip to Content
Merck
All Photos(1)

Documents

Z801658

Hellma® calibration standard Holmium Oxide glass filter F1

for testing the wavelength accuracy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.32

grade

certified reference material

manufacturer/tradename

Hellma 666-F1

application(s)

food and beverages
petroleum
pharmaceutical
plastics

format

neat

General description

Holmium oxide glass (peak positions in nm: 279; 361; 454; 536; 638)

Application

Hellma® calibration standard Holmium Oxide glass filter F1 for checking the wavelength accuracy of spectrophotometers in the UV and visible range.

Features and Benefits

  • Accredited to DAkkS Deutsche Akkreditierungsstelle (National accreditation body for the Federal Republic of Germany)
  • Traceable to NIST (National Institute of Standards and Technology)
  • Assured Quality of the measurement results
  • Proven accuracy and reliability

Other Notes

The most current version of Hellma's handling guidelines for calibration standards can be accessed on the Hellma Analytics web site.

Legal Information

Hellma is a registered trademark of Hellma GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service