Skip to Content
Merck
All Photos(1)

Key Documents

Y0001106

Medronic acid impurity B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Tetraisopropyl methylenediphosphonate

Sign Into View Organizational & Contract Pricing

Select a Size

40 μL
1.060,00 kr.

1.060,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
40 μL
1.060,00 kr.

About This Item

Linear Formula:
CH2[P(O)[OCH(CH3)2]2]2
CAS Number:
Molecular Weight:
344.32
Beilstein:
1080180
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

1.060,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

medronic acid

manufacturer/tradename

EDQM

refractive index

n20/D 1.431 (lit.)

bp

155 °C/0.5 mmHg (lit.)

density

1.08 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC(C)OP(=O)(CP(=O)(OC(C)C)OC(C)C)OC(C)C

InChI

1S/C13H30O6P2/c1-10(2)16-20(14,17-11(3)4)9-21(15,18-12(5)6)19-13(7)8/h10-13H,9H2,1-8H3

InChI key

ODTQUKVFOLFLIQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Medronic acid impurity B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service