Skip to Content
Merck
All Photos(1)

Documents

S9384

Supelco

Suloctidil

analytical standard, for drug analysis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H35NOS
CAS Number:
Molecular Weight:
337.56
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCN[C@H](C)[C@@H](O)c1ccc(SC(C)C)cc1

InChI

1S/C20H35NOS/c1-5-6-7-8-9-10-15-21-17(4)20(22)18-11-13-19(14-12-18)23-16(2)3/h11-14,16-17,20-22H,5-10,15H2,1-4H3/t17-,20-/m1/s1

InChI key

BFCDFTHTSVTWOG-YLJYHZDGSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F De Paermentier et al.
Life sciences, 44(1), 41-47 (1989-01-01)
A "cavity" lesion made by aspiration in the rat occipital cortex induces a parenchymal and a vascular reaction in its vicinity. The first was mainly characterized by cellular necrosis and gliosis, the second by an increase of the vascular network.
F Bucchi et al.
Haemostasis, 16(5), 362-368 (1986-01-01)
Oral administration of 100 or 200 mg suloctidil to healthy volunteers resulted in serum thromboxane B2 (TxB2) inhibition. This reached a maximum level between 90 min and 4 h after drug ingestion. TxB2 levels returned to 75% of basal values
Beini Zeng et al.
Oncotarget, 8(41), 69972-69982 (2017-10-21)
As the most frequent fungal pathogen in humans
G Lapathitis et al.
Biochemistry and molecular biology international, 46(5), 973-978 (1998-12-23)
Three wild-type strains of Saccharomyces cerevisiae, viz. K, Y55 and sigma 1278b, two mutants lacking one or both of the putative K+ transporters, trk1 delta and trk1 delta trk2 delta, and a mutant in the plasma membrane H(+)-ATPase, viz. pma1-105
D Thines-Sempoux et al.
Archives of toxicology, 59(2), 115-120 (1986-07-01)
The effect of suloctidil (120 mg/kg body weight PO for 3 weeks) on rat liver was investigated using biochemical and morphological methods: enzymatic activities characteristic of the main cellular compartments were used as biochemical markers of hepatocyte function and morphometry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service