PHR1688
Chlorpheniramine Related Compound C
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
3-(4-Chlorophenyl)-N-methyl-3-(pyridin-2-yl)propan-1-amine maleate salt, (3-(4-chlorophenyl)-N-methyl-3-(pyridin-2-yl)propan-1-amine maleate)
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to Ph. Eur. Y0000437
traceable to USP 1123146
API family
chlorpheniramine
CofA
current certificate can be downloaded
packaging
pkg of 50 mg
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
InChI
1S/C15H17ClN2.C4H4O4/c1-17-11-9-14(15-4-2-3-10-18-15)12-5-7-13(16)8-6-12;5-3(6)1-2-4(7)8/h2-8,10,14,17H,9,11H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
InChI key
MGNXGZAZRYTQMW-BTJKTKAUSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
Recommended products
related product
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service