Skip to Content
Merck
All Photos(1)

Key Documents

NISTRM8540

IAEA-CH-7 (carbon and hydrogenIsotopes in polyethlene foil)

NIST® RM 8540

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

isotopic purity

 2H
 13C

manufacturer/tradename

NIST®

format

matrix material

Other Notes

Reference/Informational value is provided for the following analytes.
Carbon (13C), Hydrogen (2H)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service