Key Documents
NIST8693
PFAS in aqueous film-forming foams (AFFF) formulation IV
new
NIST® SRM®
About This Item
UNSPSC Code:
12352200
format
mixture
General description
This Reference Material (RM) is intended for the identification and quantification of per- and polyfluoroalkyl substances (PFAS) in aqueous film-forming foams (AFFFs). SRM 8693_cert SRM 8693 _SDS
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Choose from one of the most recent versions:
Certificates of Analysis (COA)