NIST2942
Relative intensity correction standard for fluorescence spectroscopy
NIST® SRM® 2942, ultraviolet emission
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
solid
packaging
pkg of each (12.5 x 12.5 x 45) mm
manufacturer/tradename
NIST®
application(s)
general analytical
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Other Notes
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 1B Inhalation
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service