Skip to Content
Merck
All Photos(1)

Key Documents

NIST199

Silica brick

new

NIST SRM 199

Sign Into View Organizational & Contract Pricing

Select a Size

NIST199
4.920,00 kr.

4.920,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
NIST199
4.920,00 kr.

About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

4.920,00 kr.


Please contact Customer Service for Availability

Quality Level

packaging

pkg of 45 g

manufacturer/tradename

NIST®

application(s)

pharmaceutical (small molecule)

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) consist of approximately 45 g of silica brick powder. SRM 199_cert SRM 199 _SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service