Skip to Content
Merck
All Photos(1)

Documents

NIST1884B

Portland cement

NIST® SRM® 1884b

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 vials x 4.5 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended primarily for the evaluation or calibration of methods for analysis of cements and materials of similar matrix. A unit of SRM 1884b consists of five sealed vials, each containing approximately 4.5 g of portland cement ground to pass through a 75 μm (No. 200) sieve. For more information, please refer to the COA and SDS.

SRM 1884B_cert

SRM 1884B_SDS

Other Notes

Certified for the analytes listed below.
Silicon dioxide (SiO2), Aluminium oxide (Al2O3), Ferric oxide (Fe2O3), Calcium oxide (CaO), Magnesium oxide (MgO), Sulphur Trioxide (SO3), Sodium oxide (Na2O), Potassium oxide (K2O), Titanium dioxide (TiO2), Phosphorus pentoxide (P2O5), Manganese(III) oxide (Mn2O3), Chromium(III) oxide (Cr2O3), Strontium oxide (SrO)

Reference/Informational value is provided for the following analytes.
Zinc oxide (ZnO), Chloride (Cl), Sulfide Sulfur (S2-), Free Calcium oxide (CaO), Fluoride (F)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service