Skip to Content
Merck
All Photos(1)

Key Documents

NIST1565

Mycotoxins in Corn

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
Product NIST1565 is currently not available for purchase. Contact Technical Service

form

powder

packaging

pkg of 2 x 60 g

manufacturer/tradename

NIST®

technique(s)

NMR: suitable

application(s)

food and beverages

storage temp.

15-25°C

General description

This Standard Reference Material (SRM) is intended for validation of methods for determining mycotoxinsin corn and similar materials. A unit of SRM 1565 consists of one bottle each of two materials: low level (blank) corn and incurred corn with native levels of mycotoxins. Each bottle contains approximately 60 g of ground corn.

SRM 1565_cert

SRM 1565_SDS

Other Notes

Certified for the analytes listed below.
Deoxynivalenol, Ochratoxin A, Zearalenone

Reference/Informational value is provided for the following analytes.
Aflatoxin B1, Aflatoxin B2, Aflatoxin G1, Aflatoxin G2, Total Aflatoxins, Fumonisin B1, Fumonisin B2, Fumonisin B3, Total Fumonisins, HT-2 Toxin, T-2 Toxin

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service