Skip to Content
Merck
All Photos(1)

Key Documents

N121

Supelco

Normorphine

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17NO3
CAS Number:
Molecular Weight:
271.31
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

drug control

USDEA Schedule I; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

O[C@H]1C=C[C@H]2[C@H]3Cc4ccc(O)c5O[C@@H]1[C@]2(CCN3)c45

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Hasselström et al.
Pharmacology & toxicology, 79(1), 40-46 (1996-07-01)
Morphine, morphine-6-glucuronide and normorphine were administered to male Sprague-Dawley-rats. Analgesic effect was estimated with the hot plate and spinal nociceptive reflex depression. After intraperitoneal administration the molar potency ratio of morphine-6-glucuronide/morphine was 1.7 estimated by the paw lick latency on
M Pawula et al.
Journal of pharmaceutical and biomedical analysis, 11(4-5), 401-406 (1993-04-01)
A new, simple and rapid extraction procedure coupled with a combined coulometric-fluorescence HPLC assay is described for the simultaneous determination of morphine (M) and morphine-3-glucuronide (M3G), morphine-6-glucuronide (M6G), and normorphine (NM) in plasma. The effect of concentration and pH of
A A Coutts et al.
Canadian journal of physiology and pharmacology, 76(3), 340-346 (1998-07-23)
Cannabinoid receptor agonists inhibit electrically evoked isometric contractions of the myenteric plexus--longitudinal muscle preparation of the guinea-pig small intestine (MPLM), probably by reducing release of acetylcholine (ACh) through the activation of prejunctional CB1 receptors. As CB1 receptors are thought to
O Dale et al.
European journal of clinical pharmacology, 63(9), 837-842 (2007-07-11)
To compare the time course of morphine and metabolite concentrations in serum and cerebrospinal fluid (CSF) after intravenous and intramuscular administration after surgery. This was a randomized double-blind, double-dummy study in patients who had undergone hip replacement surgery. Morphine (M
M Zheng et al.
Journal of pharmaceutical and biomedical analysis, 16(6), 971-980 (1998-04-21)
A high-performance liquid chromatography tandem mass spectrometry-mass spectrometry (LC-MS-MS) assay was developed for the analyses of morphine, morphine glucuronides and normorphine in plasma samples from rats. The analytes were extracted by using C2 solid-phase extraction cartridges. The extraction recoveries were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service