Skip to Content
Merck
All Photos(1)

Key Documents

N1000000

Nitrosotriaminopyrimidine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2,4,6-Triamino-5-nitrosopyrimidine

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
853,00 kr.

853,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 MG
853,00 kr.

About This Item

Empirical Formula (Hill Notation):
C4H6N6O
CAS Number:
Molecular Weight:
154.13
UNSPSC Code:
41116107
NACRES:
NA.24

853,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

triamterene

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Nc1nc(c(c(n1)N)N=O)N

InChI

1S/C4H6N6O/c5-2-1(10-11)3(6)9-4(7)8-2/h(H6,5,6,7,8,9)

InChI key

XLQQJSWJHHKLOK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nitrosotriaminopyrimidine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C L Meng et al.
Nutrition and cancer, 14(3-4), 207-217 (1990-01-01)
The inhibitory effect of topical garlic extract on 7,12-dimethylbenz[a]anthracene (DMBA)-induced oral carcinogenesis in Syrian hamsters was studied to define the local anticarcinogenicity of garlic. Two other chemicals, benzo[a]pyrene (BP) and 5-nitroso-2,4,6-triaminopyrimidine (NTP), were also testing materials. All chemicals were applied

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service