Key Documents
GX308083068
Supelco
Semi Demountable Torch
for Agilent 4500/7500
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
quality
for Agilent 4500/7500
manufacturer/tradename
(30-808-3068)
compatibility
Agilent/HP 4500
Agilent/HP 7500
General description
Includes:
  • 31-808-3051 (Quartz Torch Body for Agilent SDT)
  • 31-808-0773 (Tapered Quartz Injector 2.4mm)
  • 31-808-3067 (Torch Adaptor)
  • GAZ-0604 (GazFit Connectors, 2 for 6mm OD side arms, 2 for 4mm OD side arms (PKT.4))
  • 31-100-0106 (Extractor Tool for Semi Demountable Torch, 14.2mm (standard size))
Choose from one of the most recent versions:
Certificates of Analysis (COA)