Key Documents
GX308070516
Supelco
Quartz Torch
with 1.0 mm Injector
About This Item
UNSPSC Code:
41115710
NACRES:
SB.32
quality
for Agilent7500
with 1.0 mm Injector
manufacturer/tradename
(30-807-0516)
compatibility
Agilent/HP 7500
General description
Choose GAZ-0604 GazFit Connectors for easy connection of gas tubes to torch side arms
Choose from one of the most recent versions:
Certificates of Analysis (COA)