Skip to Content
Merck
All Photos(1)

Key Documents

FA10011

Sigma-Aldrich

PIPETMAN® P200L Multichannel Micropipette

for use with BioControl® GDS, suitable for PCR, tip volume 20-200 μL

Synonym(s):

PIPETMAN P200L Multichannel Micropipette, GDS-compatible multichannel micropipette, Multichannel pipette for GDS system

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
10.840,00 kr.

10.840,00 kr.


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EACH
10.840,00 kr.

About This Item

UNSPSC Code:
41121521
NACRES:
NB.01

10.840,00 kr.


Please contact Customer Service for Availability

Request more information

packaging

box of 1 unit

technique(s)

PCR: suitable

tip volume

20-200 μL

compatibility

for use with BioControl® GDS

storage temp.

room temp

General description

PIPETMAN® P200L multichannel micropipette is a versatile laboratory tool that can be used in conjunction with the Assurance GDS® (Genetic Detection System). Assurance GDS® is an automated nucleic acid amplification system for the detection of pathogens in food, ingredients, and environmental samples. The P200L multichannel micropipette enables efficient and accurate dispensing of liquid samples across multiple wells or tubes. The pipette′s multichannel capability allows simultaneous pipetting, saving time and increasing throughput during liquid handling tasks. The PIPETMAN® P200L is equipped with advanced features such as adjustable volume settings, ergonomic design, and reliable performance, ensuring precise and reproducible dispensing of liquid samples. The compatibility of the P200L with a wide range of disposable tips further enhances the usability and adaptability of the system.

Application

PIPETMAN® P200L Multichannel Micropipette is used with Assurance GDS® to complete reagent preparation for PCR detection of pathogenic organisms in foods, ingredients, and environmental samples.

Components

The Product Contains the Following:
  • 8-channel micropipette

Legal Information

Assurance GDS is a registered trademark of BioControl Systems, Inc.
BioControl is a registered trademark of BioControl Systems, Inc.
Pipetman is a registered trademark of Gilson S.A.S. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service