Skip to Content
Merck
All Photos(1)

Key Documents

BP771

Dantrolene sodium

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

Dantrolene
sodium salt hydrate sodium salt hemi(heptahydrate), 1-[(5­(p-Nitrophenyl)furfurylidene)amino]hydantoin sodium salt hemi(heptahydrate)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
1.850,00 kr.

1.850,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
1.850,00 kr.

About This Item

Empirical Formula (Hill Notation):
C14H9N4NaO5 · 3.5H2O
CAS Number:
Molecular Weight:
399.29
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

1.850,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

dantrolene

form

solid

quality

British Pharmacopoeia (BP) Reference Standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[N+](=O)([O-])c4ccc(cc4)c5[o]c(cc5)\C=N\N6CC(=O)[N-]C6=O.[N+](=O)([O-])c1ccc(cc1)c2[o]c(cc2)\C=N\N3CC(=O)[N-]C3=O.O.O.O.O.O.O.O

InChI

1S/2C14H10N4O5.2Na.7H2O/c2*19-13-8-17(14(20)16-13)15-7-11-5-6-12(23-11)9-1-3-10(4-2-9)18(21)22;;;;;;;;;/h2*1-7H,8H2,(H,16,19,20);;;7*1H2/q;;2*+1;;;;;;;/p-2/b2*15-7+;;;;;;;;;

InChI key

LTWQNYPDAUSXBC-CDJGKPBYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dantrolene sodium BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Dantrolene Oral Suspension
  • Dantrolene Sodium
  • Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service