Skip to Content
Merck
All Photos(1)

Key Documents

BCR444

Porcine muscle (chloramphenicol blank)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

7 G
1.170,00 kr.

1.170,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
7 G
1.170,00 kr.

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

1.170,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

agriculture
cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

matrix material

storage temp.

−20°C

General description

Certified for the mass fraction of chloramphenicol.

Analysis Note

For more information please see:
BCR444

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Antibiotics: Chloramphenicol

Matrix Group: Meat
Sales restrictions may apply

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service