Skip to Content
Merck
All Photos(1)

Key Documents

BCR318

Titanium (H)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

pharmaceutical (small molecule)

format

matrix material

Packaging

100 discs

Analysis Note

For more information please see:
BCR318

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service