Skip to Content
Merck
All Photos(1)

Key Documents

BCR312

2-Nitro-7-methoxynaphtho[2,1-b]furan

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
1.170,00 kr.

1.170,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
1.170,00 kr.

About This Item

Empirical Formula (Hill Notation):
C13H9NO4
CAS Number:
Molecular Weight:
243.21
Beilstein:
5749492
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

1.170,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc2c(ccc3oc(cc23)[N+]([O-])=O)c1

InChI

1S/C13H9NO4/c1-17-9-3-4-10-8(6-9)2-5-12-11(10)7-13(18-12)14(15)16/h2-7H,1H3

InChI key

BRRIJZQYTOFDAF-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR312

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Quillardet et al.
Mutation research, 422(2), 237-245 (1998-12-05)
DNA adducts that block replication, induced in vivo by the 5-nitrofuran derivative R7000 (7-methoxy-2-nitronaphtho[2, 1-b]-furan) were mapped, at nucleotide resolution, in a region of the lacI gene of Escherichia coli, using a reiterative primer extension assay [D. Chandrasekhar, B. Van
P Quillardet et al.
Mutation research, 358(1), 113-122 (1996-10-28)
The influence of the uvr-dependent excision repair system on the lethal action, mutagenic specificity, SOS induction and DNA adducts formation of 7-methoxy-2-nitronaphtho[2,1-b]furan (R7000), a potent genotoxic nitrofuran, were examined in Escherichia coli. Binding measurements of 3H-labelled R7000 to DNA indicated
E Touati et al.
Mutagenesis, 4(4), 254-258 (1989-07-01)
The effects on DNA, in bacteria, of 7-methoxy-2-nitro-naphtho[2,1-b]furan (R7000), a very potent genotoxic product from the 2-nitronaphthofuran series, were investigated with two different approaches: (i) measurement of the binding of the radiolabelled mutagen to DNA and (ii) detection by the
E Touati et al.
Carcinogenesis, 17(12), 2543-2550 (1996-12-01)
The mutagenic specificity of 7-methoxy-2-nitronaphtho[2,1-b]furan (R7000), a very potent genotoxic 2-nitrofuran, was investigated in the lacI gene of E.coli. To analyze the influence of SOS-mutagenesis on R7000-induced mutations, 86 and 84 LacI- mutants were respectively isolated from umuC+ and umuC
P Quillardet et al.
Mutation research, 204(2), 141-147 (1988-02-01)
We measured the genotoxic activities in two bacterial tests, the Salmonella/histidine assay (a reverse mutation assay) and the SOS chromotest (an assay for SOS induction in E. coli), of three 2-nitroanthrafurans: 2-nitroanthra[1,2-b]furan (R-7688), the isomeric compound 2-nitroanthra[2,1-b]furan (R-7686) and its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service