Skip to Content
Merck
All Photos(1)

Documents

B7181

Sigma-Aldrich

N-(+)-Biotinyl-6-aminohexanoic acid

solid

Synonym(s):

(+)-Biotin-ε-aminocaproic acid, 6-((Biotinoyl)amino)hexanoic acid, Biotin-X, N-(+)-Biotinyl-6-aminocaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H27N3O4S
CAS Number:
Molecular Weight:
357.47
Beilstein:
706392
MDL number:
UNSPSC Code:
12352200

form

solid

solubility

DMF: soluble (pH>6)

SMILES string

[H][C@]12CS[C@@H](CCCCC(=O)NCCCCCC(O)=O)[C@@]1([H])NC(=O)N2

InChI

1S/C16H27N3O4S/c20-13(17-9-5-1-2-8-14(21)22)7-4-3-6-12-15-11(10-24-12)18-16(23)19-15/h11-12,15H,1-10H2,(H,17,20)(H,21,22)(H2,18,19,23)/t11-,12-,15-/m0/s1

InChI key

CMUGHZFPFWNUQT-HUBLWGQQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Biotinylation reagent

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service