Skip to Content
Merck
All Photos(2)

Documents

A0223

Sigma-Aldrich

3-Amino-3-deoxydigoxigenin hemisuccinamide, succinimidyl ester

≥80% purity (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

3-Amino-3-deoxydigoxigenin hemisuccinamide, succinimidyl ester,

Assay

≥80% (HPLC)

Quality Level

form

solid

mol wt

586.68

color

white

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@]12CC[C@@H](C[C@H]1CC[C@@H]3[C@@H]2C[C@@H](O)[C@]4(C)[C@H](CC[C@]34O)C5=CC(=O)OC5)NC(=O)CCC(=O)ON6C(=O)CCC6=O

InChI

1S/C31H42N2O9/c1-29-11-9-19(32-24(35)5-8-27(38)42-33-25(36)6-7-26(33)37)14-18(29)3-4-21-22(29)15-23(34)30(2)20(10-12-31(21,30)40)17-13-28(39)41-16-17/h13,18-23,34,40H,3-12,14-16H2,1-2H3,(H,32,35)/t18-,19+,20-,21-,22+,23-,29+,30+,31+/m1/s1

InChI key

AVQGRLJRQILBRQ-ZCIIQETRSA-N

Application

Amine-reactive haptenylation reagent. This succinimidyl ester derivative of digoxigenin has been shown to inhibit the Na+/K+- ATPase by binding to the cardiac steroid receptor site.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service