Skip to Content
Merck
All Photos(1)

Key Documents

99697

Supelco

Silica gel C18 on TLC Plates

chiral modified, impregnated with Cu2+ reagent, chiral-silica gel matrix, L × W 5 cm × 20 cm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

material

chiral-silica gel matrix
glass support

quality

chiral modified
impregnated with Cu2+ reagent

feature

binder Organic
fluorescent indicator

packaging

pkg of 50 ea

manufacturer/tradename

Sigma-Aldrich

technique(s)

thin layer chromatography (TLC): suitable

L × W

5 cm × 20 cm

layer thickness

250 μm

particle size

≥8 μm

pore size

60 Å medium pore diameter

Looking for similar products? Visit Product Comparison Guide

General description

C18 possess 18 carbon atoms in the acyl chain, commonly used as chelating agent, is highly reactive, stable and hydrophobic in nature. It can be used as an effective metal extraction agent. C18 supported on silica gel surface can withstand large volumes of metal ion solution without a releasing problem.[1]

Application

Silica gel C18 on TLC Plates may be used in thin layer chromatography and HPTLC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pre-concentration of copper, cobalt and nickel with 3-methyl-1-phenyl-4-stearoyl-5-pyrazolone loaded on silica gel
Tong, A., Yoshifumi Akama, and S. Tanaka.
Analyst, 115, 947-949 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service