Skip to Content
Merck
All Photos(1)

Key Documents

93169

Supelco

Eicosanyl caffeate

analytical standard

Synonym(s):

Caffeic acid eicosyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H48O4
CAS Number:
Molecular Weight:
460.69
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

CCCCCCCCCCCCCCCCCCCCOC(=O)\C=C\c1ccc(O)c(O)c1

InChI

1S/C29H48O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-24-33-29(32)23-21-26-20-22-27(30)28(31)25-26/h20-23,25,30-31H,2-19,24H2,1H3/b23-21+

InChI key

LTSJTDDQUOUKJT-XTQSDGFTSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

354.2 °F - closed cup

Flash Point(C)

179 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service