Skip to Content
Merck
All Photos(1)

Key Documents

85515

Millipore

Sorbic acid Agar (Base)

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
Pricing and availability is not currently available.

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 16 g/L
casein peptone, 10 g/L
D(+)-glucose, 20 g/L
magnesium sulfate heptahydrate, 0.2 g/L
manganous sulfate, 0.05 g/L
meat extract, 10 g/L
sodium acetate, 5 g/L
sodium citrate, 3 g/L
yeast extract, 5 g/L

final pH

5.0±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages

microbiology

suitability

selective by low pH for Lactobacillus spp.

Application

For the isolation and differentiation of lactobacilli from food, faeces etc. according to Reuter.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Reuter
Fleischwirtsch., 954-954 (1970)
G. Reuter
Archiv Fur Lebensmittelhygiene, 19, 53-53 (1968)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service