Skip to Content
Merck
All Photos(1)

Documents

82570

Sigma-Aldrich

(+)-Pulegone

purum, ≥95.0% (sum of enantiomers, GC)

Synonym(s):

(R)-2-Isopropylidene-5-methylcyclohexanone, (R)-p-Menth-4(8)-en-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
2040703
EC Number:
MDL number:
UNSPSC Code:
12352000

grade

purum

Assay

≥95.0% (sum of enantiomers, GC)

optical activity

[α]20/D +23±1.5°, neat

refractive index

n20/D 1.487

bp

223-224 °C (lit.)

density

0.935 g/mL at 20 °C (lit.)

SMILES string

C[C@@H]1CCC(=C(\C)C)\C(=O)C1

InChI

1S/C10H16O/c1-7(2)9-5-4-8(3)6-10(9)11/h8H,4-6H2,1-3H3/t8-/m1/s1

InChI key

NZGWDASTMWDZIW-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Chiral building block Synthesis of (-)-8-phenylmenthol

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.D. White et al.
Tetrahedron Letters, 28, 3061-3061 (1987)
T. Zair et al.
The Journal of Organic Chemistry, 58, 2686-2686 (1993)
M. Alami et al.
Organic Syntheses, 72, 135-135 (1995)
O. Ort
Organic Syntheses, 65, 203-203 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service