Skip to Content
Merck
All Photos(1)

Documents

80312

Supelco

Perfluorotetradecanoic acid

analytical standard

Synonym(s):

PFAS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)12CO2H
CAS Number:
Molecular Weight:
714.11
EC Number:
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for EPA 1633
suitable for EPA 537.1
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45
suitable for ISO 21675 2019

Assay

≥96.0% (GC)
96.0-104.0% (T)

shelf life

limited shelf life, expiry date on the label

bp

270 °C/740 mmHg (lit.)

mp

130-135 °C (lit.)

application(s)

environmental

format

neat

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C14HF27O2/c15-2(16,1(42)43)3(17,18)4(19,20)5(21,22)6(23,24)7(25,26)8(27,28)9(29,30)10(31,32)11(33,34)12(35,36)13(37,38)14(39,40)41/h(H,42,43)

InChI key

RUDINRUXCKIXAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Perfluorotetradecanoic acid belongs to the class of perfluorinated compounds, which are found to be persistent in the environment with bioaccumulative potential. It is used in various commercial applications, including firefighting foams, detergents, insecticides, paper, food containers, as well as to produce photographic films and manufacture fat- and wate-resistant fabrics, etc.

Application

Perfluorotetradecanoic acid may be used as an analytical reference standard for the quantification of the analyte in water samples and Anguilla anguilla using high-performance liquid chromatography technique.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Detection of perfluoroalkyl acids and sulphonates in Italian eel samples by HPLC-HRMS Orbitrap
Chiesa ML, et al.
Chemosphere, 193, 358-364 (2018)
Perfluorinated compounds in the Pearl river and Yangtze river of China
So KM, et al.
Chemosphere, 68(11), 2085-2095 (2007)

Articles

Liquid Chromatography–Mass Spectrometry (LC-MS) separation of 33 PFAS (Per- and poly-fluoroalkyl substances) using a superficially porous silica particles (SPP) based Ascentis® Express C18 column.

Find more about the analysis of 16 PFAS in salmon via QuEChERS extraction and LC-MS/MS characterization following FDA Method C-010.02.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service