Skip to Content
Merck
All Photos(1)

Key Documents

80180

Sigma-Aldrich

Phytic acid solution

technical, ~40% in H2O

Synonym(s):

myo-Inositol hexakis(dihydrogen phosphate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H18O24P6
CAS Number:
Molecular Weight:
660.04
Beilstein:
2201952
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

technical

form

solution

concentration

~40% in H2O

refractive index

n20/D 1.39

SMILES string

OP(O)(=O)O[C@@H]1[C@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@H]1OP(O)(O)=O

InChI

1S/C6H18O24P6/c7-31(8,9)25-1-2(26-32(10,11)12)4(28-34(16,17)18)6(30-36(22,23)24)5(29-35(19,20)21)3(1)27-33(13,14)15/h1-6H,(H2,7,8,9)(H2,10,11,12)(H2,13,14,15)(H2,16,17,18)(H2,19,20,21)(H2,22,23,24)/t1-,2-,3-,4+,5-,6-

InChI key

IMQLKJBTEOYOSI-GPIVLXJGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Complexing agent for the removal of traces of heavy metal ions

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenbo Zhao et al.
Biosensors & bioelectronics, 44, 1-5 (2013-02-05)
In this work, the silica- phytic acid (SiO2-PA) nanocomposites were synthesized by the method of reverse microemulsion and electrostatic binding. The newly designed materials were used to develop a novel glucose biosensor by immobilizing glucose oxidase (GOx) onto the SiO2-PA
Beata Parfiniewicz et al.
Acta poloniae pharmaceutica, 69(6), 1291-1297 (2013-01-05)
The latest studies suggest that adhesion molecules are involved in the arising of malignant changes and in distant metastasis induction. The soluble forms of several adhesion molecules, have recently emerged as novel and potentially useful tumor markers. Among a number
Rajeswari Ravichandran et al.
Macromolecular bioscience, 13(3), 366-375 (2013-01-22)
A functional scaffold fabricated is developed from natural polymers, favoring regeneration of the ischemic myocardium. Hemoglobin/gelatin/fibrinogen (Hb/gel/fib) nanofibers are fabricated by electrospinning and are characterized for morphology, scaffold composition, functional groups and hydrophilicity. It is hypothesized that ex vivo pretreatment
Siwaporn Pinkaew et al.
The Journal of nutrition, 143(3), 362-368 (2013-01-11)
Iron (Fe), zinc (Zn), and vitamin A (VA) deficiencies are common among children in developing countries and often occur in the same individual. Rice is widely consumed in the developing countries of Asia and the low phytate in polished rice
Christopher J Barker et al.
Pharmacological reviews, 65(2), 641-669 (2013-02-23)
Studies of inositol polyphosphates in the pancreatic β-cell have led to an exciting synergism between new discoveries regarding their cellular roles and new insights into β-cell function. Because the loss or malfunction of the β-cell is central to diabetes, these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service