Skip to Content
Merck
All Photos(1)

Documents

74190

Sigma-Aldrich

Nonadecanoic acid

≥98.0% (GC)

Synonym(s):

Nonadecylic acid, C19:0, NSC 11914

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17COOH
CAS Number:
Molecular Weight:
298.50
Beilstein:
1786261
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥98.0% (GC)

form

powder or crystals

mp

68-70 °C (lit.)

functional group

carboxylic acid

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C19H38O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21/h2-18H2,1H3,(H,20,21)

InChI key

ISYWECDDZWTKFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Nonadecanoic acid, a 19-carbon saturated fatty acid, has anti-tumor activity. It has inhibitory activity on the proliferation of HL-60 cells, a human tumor cell line .

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masataka Fukuzawa et al.
Biological & pharmaceutical bulletin, 31(10), 1933-1937 (2008-10-02)
During our isolation of biologically active substances from the spores of Ganoderma lucidum (Reishi Houshi, G. lucidum) guided by the inhibitory activity on HL-60 cell proliferation, NMR spectroscopic and mass spectrometric data indicate the substance contains a mixture of several
Bhushan V Kulkarni et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 306(12), R879-R885 (2014-04-04)
Lingual lipase generates nonesterified fatty acids (NEFA) from dietary fats during oral processing by lipolysis. Lingual lipase in rodents has strong lipolytic activity and plays a critical role in oral detection of fats. The functional activity of lingual lipase during
Alok Patel et al.
Bioresource technology, 165, 214-222 (2014-04-22)
Hemp seeds aqueous extract (HSAE) was used as cheap renewable feedstocks to grow novel oleaginous yeast Rhodosporidium kratochvilovae HIMPA1 isolated from Himalayan permafrost soil. The yeast showed boosted triglyceride (TAG) accumulation in the lipid droplets (LDs) which were transesterified to
Ji Hyung Kim et al.
Journal of microbiology and biotechnology, 25(2), 238-246 (2014-11-11)
A unicellular red microalga was isolated from environmental freshwater in Korea, and its morphological, molecular, and biochemical properties were characterized. Morphological analysis revealed that the isolate was a unicellular biflagellated green microalga that formed a non-motile, thick-walled palmelloid or red
L J Qu et al.
Journal of insect science (Online), 14, 166-166 (2014-11-05)
Radiation-induced sterile insect technique is a biologically based, environment-friendly method for the suppression or eradication of a number of insect pests. Although the basic mechanisms underlying the technology have been well studied, little is known about the cell responses in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service