Skip to Content
Merck
All Photos(2)

Documents

73348

Supelco

N-Desmethylthiamethoxam

PESTANAL®, analytical standard

Synonym(s):

3-[(2-Chloro-1,3-thiazol-5-yl)methyl]-N-nitro-1,3,5-oxadiazinan-4-imine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8ClN5O3S
CAS Number:
Molecular Weight:
277.69
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+](=O)N=C1NCOCN1Cc2cnc(Cl)s2

InChI

1S/C7H8ClN5O3S/c8-6-9-1-5(17-6)2-12-4-16-3-10-7(12)11-13(14)15/h1H,2-4H2,(H,10,11)

InChI key

LOXCNVOJGRNBFX-UHFFFAOYSA-N

General description

N-Desmethylthiamethoxam belongs to the class of neonicotinoid insecticides, widely used in the control of Colorado potato beetle, a major pest in potato cultivation.
Find more information here: Neonicotinoids

Application

N-Desmethylthiamethoxam may be used as an analytical reference standard for the quantification of the analyte in wine samples using liquid chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colorado potato beetle resistance to insecticides
Alyokhin A, et al.
American Journal of Potato Research, 85(6), 395-413 (2008)
Adela Jing Li et al.
Environment international, 135, 105415-105415 (2019-12-24)
Human exposure to neonicotinoid insecticides (hereafter "neonics") is a concern. Spot urine samples have been widely used in the assessment of exposure to neonics. Urinary concentrations, however, can vary greatly over time due to variable exposure, potentially leading to exposure
J D Berset et al.
Journal of chromatography. A, 1506, 45-54 (2017-05-28)
A direct large volume injection (DI-LVI) high performance liquid chromatography - tandem mass spectrometry (HPLC-MS/MS) method was developed and validated for the quantitative determination of 16 systemic insecticides and their main plant metabolites. The assays were conducted on commercial red

Protocols

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Analysis of banned neonicotinoid insecticides from dandelion blossoms using QuEChERS and LC-MS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service