Skip to Content
Merck
All Photos(1)

Documents

70660

Sigma-Aldrich

2-Naphthoxyacetic acid

technical, ≥90% (T)

Synonym(s):

BNOA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OCH2CO2H
CAS Number:
Molecular Weight:
202.21
Beilstein:
1074148
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

technical

Assay

≥90% (T)

mp

151-154 °C (lit.)
151-154 °C

SMILES string

OC(=O)COc1ccc2ccccc2c1

InChI

1S/C12H10O3/c13-12(14)8-15-11-6-5-9-3-1-2-4-10(9)7-11/h1-7H,8H2,(H,13,14)

InChI key

RZCJYMOBWVJQGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Kokkinou et al.
Carbohydrate research, 332(1), 85-94 (2001-06-14)
The structure of the complex of beta-cyclodextrin (cyclomaltoheptaose) with beta-naphthyloxyacetic acid was studied in solid state by X-ray diffraction and in aqueous solution by 1H NMR spectroscopy. The complex crystallizes in the channel mode, space group C2, with a stoichiometry
L Li et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 56A(8), 1513-1521 (2000-07-25)
A cyclodextrin induced room temperature phosphorimetry (CD-RTP) for determine beta-NOA, which using I- as a heavy atom perturber (HAP) and sodium sulfite as a deoxygenator, was developed. The phosphorescence peak wavelength maxima lambda(ex)/lambda(em) = 287/496,521 nm. The analytical curve of
Chun Zhou et al.
Environmental monitoring and assessment, 154(1-4), 233-239 (2008-06-17)
A reliable and sensitive competitive fluorescence immunoassay for the quantitative determination of naphthalene (NA) was developed. 2-naphthoxy acetic acid (NAA) was selected as the hapten of naphthalene. Active ester method (AEM) was used to couple the NAA to carrier proteins
Tsai-Hui Duh et al.
Journal of chromatography. A, 987(1-2), 205-209 (2003-03-05)
In continuing the search for fluorescent reagents for analytical derivatization in chromatography, we found a simple chemical, (2-naphthoxy)acetyl chloride, with potential fluorophore/chromophore characteristics for the highly sensitive detection of analytes with an amino function. The reagent has an auxochrome (a
Sanghamitra Atta et al.
Journal of photochemistry and photobiology. B, Biology, 111, 39-49 (2012-04-20)
We report a novel technique for controlled release of plant growth regulators (PGRs) by sunlight using photoremovable protecting group (PRPG) as a delivery device. In the present work, carboxyl-containing PGRs of the auxin group [indoleacetic acid (IAA) and naphthoxyacetic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service