Skip to Content
Merck
All Photos(1)

Key Documents

68328

Supelco

16-O-Methylcafestol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water (Karl Fischer)

application(s)

environmental
food and beverages

format

neat

SMILES string

CO[C@]1(CO)C[C@@]23CCC4c5ccoc5CC[C@@]4(C)C2CCC1C3

InChI

1S/C21H30O3/c1-19-8-6-17-15(7-10-24-17)16(19)5-9-20-11-14(3-4-18(19)20)21(12-20,13-22)23-2/h7,10,14,16,18,22H,3-6,8-9,11-13H2,1-2H3/t14-,16-,18+,19-,20+,21+/m1/s1

InChI key

BDVVNPOGDNWUOI-GVOJMRIRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

16-O-Methylcafestol, a diterpene, is present as a major constituent in robusta coffee oil.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diterpenes and diterpene esters in coffee
Kurzrock T and Speer.K
Food Reviews International, 17(4), 433-450 (2001)
Food Authentication
Ashrust.RP and Dennis.JM
Food Authentication (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service