Skip to Content
Merck
All Photos(1)

Key Documents

66010

Supelco

Hydroxycitronellal

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O2
CAS Number:
Molecular Weight:
172.26
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥94.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.447-1.450
n20/D 1.448 (lit.)

bp

257 °C (lit.)

density

0.923 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

[H]C(=O)CC(C)CCCC(C)(C)O

InChI

1S/C10H20O2/c1-9(6-8-11)5-4-7-10(2,3)12/h8-9,12H,4-7H2,1-3H3

InChI key

WPFVBOQKRVRMJB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydroxycitronellal belongs to a broad range of flavor and fragrance standards for quality control of food and cosmetic products provided by Sigma-Aldrich. It is reported as a sensitizing fragrance allergen found in many products. Hydroxycitronella is commonly used in the cosmetic industry to neutralize unpleasant odors that may be present in the chemicals, to enhance the individuality and appeal of products, fragranced cosmetics including aftershaves, eye-creams, etc.
Hydroxycitronellal is a synthetic fragrance material used in perfumes. It can be prepared from citronellal either by hydration of citronellal bisulfite or by hydrolysis with sulphuric acid and alkali.

Application

Hydroxycitronellal has been used as an analytical reference standard for the quantification of the analyte in fragrance samples using flow modulation comprehensive 2D GC/MS (FM GCxGC/MS) method.
It may also be used as an analytical reference standard for the quantification of the analyte in the following:
  • Fragrances using comprehensive two-dimensional gas chromatography coupled with rapid scanning quadrupole mass spectrometry (GC × GC-qMS).
  • Wastewater treatment plants (WWTPs) using gas chromatography-mass spectrometry (GC-MS).

It was used as reference standard for the validation of fragrance allergens in cosmetic products using solid-phase microextraction method (SPME) - gas chromatography coupled with flame ionization detector (GC–FID).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation of SPME-GC-FID Method for Determination of Fragrance Allergens in Selected Cosmetic Products.
Divisova, R., et al.
Acta Chromatographica , 27.3, 509-523 (2015)
Identification, quantitation and method validation for the analysis of suspected allergens in fragrances by comprehensive two-dimensional gas chromatography coupled with quadrupole mass spectrometry and with flame ionization detection
Cordero C, et al.
Journal of Chromatography A, 1150(1-2), 37-49 (2007)
Synthesis of hydroxycitronellal. Hydration and subsequent hydrolysis of imines, enamines, or oxazolidines prepared from citronellal and amines.
Ishino, Ritsuko, and Ju Kumanotani
The Journal of Organic Chemistry, 39.1, 108-111 null
Dictionary of Contact Allergens, 2357-2471 (2019)
Monitoring of sixteen fragrance allergens and two polycyclic musks in wastewater treatment plants by solid phase microextraction coupled to gas chromatography
Godayol A, et al.
Chemosphere, 119(1), 363-370 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service