Skip to Content
Merck
All Photos(1)

Key Documents

61074

Supelco

Propiophenone

analytical standard

Synonym(s):

Ethyl phenyl ketone

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
366,00 kr.

366,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
366,00 kr.

About This Item

Linear Formula:
C6H5COC2H5
CAS Number:
Molecular Weight:
134.18
Beilstein:
606215
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

366,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor pressure

1 mmHg ( 50 °C)

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.526 (lit.)
n20/D 1.527

bp

218 °C (lit.)

mp

17-19 °C (lit.)

density

1.009 g/mL at 20 °C
1.009 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCC(=O)c1ccccc1

InChI

1S/C9H10O/c1-2-9(10)8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

KRIOVPPHQSLHCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adriana Arigò et al.
Journal of chromatography. A, 1649, 462183-462183 (2021-05-27)
In the European Union, cosmetic products are regulated by the Cosmetics Regulation (EC) No. 1223/2009. The Analytical Working Group of the International Fragrance Association (IFRA) suggested that a new sensitive analytical method is needed to determine psoralens in finished cosmetic
Martin Gilar et al.
Journal of chromatography. A, 1390, 86-94 (2015-03-10)
Chromatographic zone broadening is a common issue in microfluidic chromatography, where the sample volume introduced on column often exceeds the column void volume. To better understand the propagation of wide chromatographic zones on a separation device, a series of MS
Martin Gilar et al.
Journal of chromatography. A, 1381, 110-117 (2015-01-22)
Sample introduction in microfluidic liquid chromatography often generates wide zones rather than peaks, especially when a large sample volume (relative to column volume) is injected. Formation of wide injection zones can be further amplified when the sample is dissolved in
Yan Ma et al.
Journal of chromatography. A, 1383, 47-57 (2015-02-05)
Prototype small-size (1.0mm I.D., 5cm long) columns for reversed-phase HPLC were evaluated in relation to instrument requirements. The performance of three types of columns, monolithic silica and particulate silica (2μm, totally porous and 2.6μm, core-shell particles) was studied in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service