60038BC
VIP® Gold Salmonella Kit
suitable for immunological and lateral flow essay testing in Environmental, and Food and Beverages applications
About This Item
Recommended Products
product name
VIP® Gold Salmonella, BioControl, Lateral flow test for detection of Salmonella in food and environmental samples
Agency
AOAC® (OMA)
AOAC® (PTM)
product line
VIP®
form
liquid
shelf life
3‑9 mo.
packaging
pkg of 40
manufacturer/tradename
BioControl Systems 60038-40
VIP®
storage condition
dry at room temperature
technique(s)
lateral flow assay: suitable
microbe id | immunological: suitable
input
food(s)
application(s)
food and beverages
pathogen testing
storage temp.
room temp
suitability
Salmonella spp.
General description
Application
- AOAC Official Method of Analysis 999.09
- AOAC Performance Tested Method 031801
Features and Benefits
- Fast Results: Next day results for processed foods; raw and highly contaminated foods require an additional overnight incubation.
- Improved Lab Efficiency: VIP Gold offers an innovative multi-test format with break-away devices designed for maximum flexibility to accommodate a wide range of testing volumes.
- Advanced Technology: The cutting edge particle technology allows for faster sample flow, producing a clean background, clear lines and easy to read positive/negative results without the need for a reader.
Components
- One foil pouch of 40 tests
- Extraction reagents
Compatibility
Testing Method
Legal Information
Kit Components Only
- Adhesive Coated Clear Polyester Backing 0.005”
- Absorbent Blotter Sheet
- VIP® Prefilter Sample Pad, Cut
- VIP® Salmonella Gold Colloid Label Pad
- VIP® Salmonella Gold Reaction Membrane
- Desiccant, 14g
also commonly purchased with this product
related product
required but not provided
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service