Skip to Content
Merck
All Photos(1)

Key Documents

58087

Supelco

di-Sodium hydrogen phosphate dihydrate

suitable for HPLC, LiChropur

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate dihydrate, Disodium phosphate, Sodium phosphate dibasic dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
274,00 kr.
250 G
866,00 kr.

274,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
274,00 kr.
250 G
866,00 kr.

About This Item

Linear Formula:
Na2HPO4 · 2H2O
CAS Number:
Molecular Weight:
177.99
EC Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

274,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98.5-102.0% (T)

manufacturer/tradename

LiChropur

technique(s)

HPLC: suitable

pH

8.8-9.4

solubility

H2O: 10%, clear, colorless

λ

10 % in H2O

UV absorption

λ: 230 nm Amax: ≤0.1
λ: 260 nm Amax: ≤0.06
λ: 280 nm Amax: ≤0.04
λ: 320 nm Amax: ≤0.02
λ: 500 nm Amax: ≤0.02

SMILES string

O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.2H2O/c;;1-5(2,3)4;;/h;;(H3,1,2,3,4);2*1H2/q2*+1;;;/p-2

InChI key

KDQPSPMLNJTZAL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service