Skip to Content
Merck
All Photos(1)

Documents

50463

Sigma-Aldrich

4,N,N-Trimethylaniline

puriss., catalyst for peroxide polymerisations, ≥99.0% (GC)

Synonym(s):

4-Dimethylaminotoluene, N,N-Dimethyl-p-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4N(CH3)2
CAS Number:
Molecular Weight:
135.21
Beilstein:
774409
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

catalyst for peroxide polymerisations
puriss.

vapor density

>1 (vs air)

Assay

≥99.0% (GC)

expl. lim.

7 %

refractive index

n20/D 1.546 (lit.)
n20/D 1.547

bp

211 °C (lit.)
90-92 °C/10 mmHg (lit.)

density

0.937 g/mL at 25 °C (lit.)

SMILES string

CN(C)c1ccc(C)cc1

InChI

1S/C9H13N/c1-8-4-6-9(7-5-8)10(2)3/h4-7H,1-3H3

InChI key

GYVGXEWAOAAJEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Caution

may discolor to yellowish-green on storage

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1B - Repr. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Reproductive organs

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Makihira et al.
Mycoses, 45(8), 300-305 (2003-02-08)
The effects of four liquid components of denture acrylic resin on host cell activity and fungal adhesion were investigated in this study. The low concentration (1 micromol l(-1)) of the liquid components caused no change in the activities and morphologies
Y Nomura et al.
Journal of materials science. Materials in medicine, 17(1), 29-32 (2006-01-04)
The polymerization initiators for resins cured using visible light usually consist of a photosensitizer, primarily camphorquinone (CQ), and a reducing agent, which is often a tertiary amine (DMPT, DMAEMA), while the initiator used for self-curing resins consists of benzoyl peroxide
M Noda et al.
Journal of biomedical materials research. Part A, 83(1), 123-129 (2007-03-27)
Resin composites are widely used in dentistry, and are polymerized in situ using a blue-light activated, free-radical polymerization mechanism. Blue light (400-500nm) is used to activate camphoroquinone (CQ), which decomposes to form free radicals that are stabilized by dimethyl-p-toludine (DMPT).
Jennifer L Moreau et al.
Journal of biomedical materials research. Part A, 81(3), 594-602 (2006-12-21)
There is an increasing need to develop new biomaterials as tissue engineering scaffolds. Unfortunately, many of the materials that have been studied for these purposes are polyesters that hydrolytically degrade into acidic products, which may harm the surrounding tissue, and
Sachiko Kaihara et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 68(1), 67-73 (2007-09-25)
While many synthetic, hydrolytically degradable hydrogels have been developed for biomedical applications, there are only a few examples whose polymer backbone does not form acidic products upon degradation. In order to address this concern, we proposed to develop a hydrogel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service