Skip to Content
Merck
All Photos(1)

Key Documents

50279

Millipore

Listeria UVM II Supplement

suitable for microbiology

Synonym(s):

Antibiotic supplement, Listeria secondary selective enrichment supplement (UVM II)

Sign Into View Organizational & Contract Pricing

Select a Size

5 VIALS
429,00 kr.

429,00 kr.


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
5 VIALS
429,00 kr.

About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

429,00 kr.


Please contact Customer Service for Availability

Request more information

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

acriflavin hydrochloride, 12.5 mg per vial
nalidixic, 10.0 mg

packaging

pkg of 5 vials

application(s)

environmental
food and beverages
pathogen testing

microbiology

storage temp.

2-8°C

suitability

Listeria spp.

General description

Listeria UVM II Supplement is used as an antibiotic supplement in culture media to inhibit the growth of unwanted organisms while allowing the growth of the desired microorganisms. This antibiotic supplement is recommended for the selective isolation and enumeration of Listeria sp. The supplement consists of a combination of Acriflavin hydrochloride (12.5 mg) and Nalidixic (10 mg) with broad-spectrum antimicrobial activity against various Gram-positive and Gram-negative organisms. thereby selectively allowing the growth of Listeria sp.

Application

Listeria UVM II Supplement is recommended for the selective isolation and enumeration of Listeria species from various food and clinical samples.

Components

Composition:
(per vial, sufficient for 500 mL medium)
Acriflavin hydrchloride 12.5 mg
Nalidixic 10.0 mg

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service