Skip to Content
Merck
All Photos(1)

Key Documents

46014

Supelco

1,2,3,4-Tetrachlorobenzene

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Cl4
CAS Number:
Molecular Weight:
215.89
Beilstein:
1910025
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

254 °C/761 mmHg (lit.)

mp

42-47 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1ccc(Cl)c(Cl)c1Cl

InChI

1S/C6H2Cl4/c7-3-1-2-4(8)6(10)5(3)9/h1-2H

InChI key

GBDZXPJXOMHESU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ken G Drouillard et al.
Bulletin of environmental contamination and toxicology, 93(5), 561-566 (2014-06-18)
A vial equilibration technique was used to estimate the fugacity capacities of food and feces samples for 1,2,3,4-tetrachlorobenzene (TCB). The method was calibrated using different volumes of n-octanol and by comparing the measured and predicted fugacity capacity (Zoct) of n-octanol
B A Kimball et al.
Journal of wildlife diseases, 32(3), 505-511 (1996-07-01)
Pentachlorobenzene (PeCB) and 1,2,3,4-tetrachlorobenzene (TeCB) were evaluated as oral chemical biomarkers when administered to coyotes (Canis latrans) during the period of 31 January to 10 August 1994. Three coyotes each received 100 mg of PeCB and three received 100 mg
L D Bowers et al.
Clinical chemistry, 43(6 Pt 1), 1033-1039 (1997-06-01)
Limits of detection are important issues for GC/MS screening for anabolic agents and for confirmation of various drugs of abuse. We compared a quadrupole ion trap (QIT) operated in two different selected-ion storage modes and a quadrupole mass filter (QMF)
Kenichiro Inoue et al.
Environmental science & technology, 39(15), 5844-5850 (2005-08-30)
Carbonaceous adsorbents such as activated carbon have been used to reduce the emission of organic pollutants from incineration plants. However, with this method, the amount and type of adsorbent to be used are based only on empirical results, which may
K T Kitchin et al.
Toxicology, 26(3-4), 243-256 (1983-03-01)
To assess possible maternal hepatic and reproductive effects of this uncharged, low molecular weight, lipophilic chlorinated benzene 0, 100, 300 and 1000 mg/kg/day of 1,2,3,4-tetrachlorobenzene (TCB) was orally administered to pregnant rats on days 9-13 of gestation and the animals

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service