Skip to Content
Merck
All Photos(1)

Documents

45349

Supelco

Bitertanol

PESTANAL®, analytical standard

Synonym(s):

3,3-Dimethyl-1-(4-phenylphenoxy)-1-(1,2,4-triazol-1-yl)butan-2-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23N3O2
CAS Number:
Molecular Weight:
337.42
Beilstein:
620948
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

mixture of stereo isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)(C)C(O)C(Oc1ccc(cc1)-c2ccccc2)n3cncn3

InChI

1S/C20H23N3O2/c1-20(2,3)18(24)19(23-14-21-13-22-23)25-17-11-9-16(10-12-17)15-7-5-4-6-8-15/h4-14,18-19,24H,1-3H3

InChI key

VGPIBGGRCVEHQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A R Allen et al.
Neurotoxicology and teratology, 15(4), 237-242 (1993-07-01)
Several recent reports indicate that triadimefon, a triazole fungicide, has effects on behavior that are similar to those of psychomotor stimulants. For example, triadimefon increases overall fixed-interval (FI) response rate, disrupts FI response patterning, increases motor activity, and produces stereotypies
Rafael Boscolo-Berto et al.
Legal medicine (Tokyo, Japan), 47, 101742-101742 (2020-07-13)
The fatal rupture of a saccular aneurysm at the junction between the left anterior cerebral artery and anterior communicating artery affected by fibromuscular dysplasia (FMD) is a rare condition. Here is reported the case of a subject involved in a
Ping-Kun Chan et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 44(12), 2047-2057 (2006-09-15)
The effects of fungicide bitertanol on cytochrome P450-dependent monooxygenases were studied using rats treated intraperitoneally with the N-substituted triazole for 4 days. Treatment with 10, 25, and 100 mg/kg bitertanol produced 2-, 4-, and 14-fold increases of 7-ethoxyresorufin O-deethylation activity
Y Yamazaki et al.
Journal of AOAC International, 81(6), 1252-1256 (1998-12-16)
A simple and rapid method was developed for determining bitertanol residues in strawberries. Bitertanol was extracted from samples with ethyl acetate. Bitertanol acetate was added prior to extraction as a surrogate standard. The ethyl acetate extract was cleaned up by
Marc Bourgin et al.
Chemosphere, 90(4), 1387-1395 (2012-09-25)
The degradation of bitertanol by ozone treatment is investigated. Solutions of bitertanol (8.4 μg mL(-1)) were prepared either by dissolution of the standard or by dilution of Gaucho Blé seed loading solution and then ozonated under different conditions. Evolution of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service